Assay Details
| Gene Symbol: Racgap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001253808.1 | NP_001240737.1 | 5 - 6 | 651 | 137 | |
| NM_001253809.1 | NP_001240738.1 | 5 - 6 | 647 | 137 | ||
| NM_012025.7 | NP_036155.1 | 6 - 7 | 782 | 137 | ||
| GenBank mRNA | AB030252.1 | - | 5 - 6 | 631 | 137 | |
| AF079974.1 | - | 5 - 6 | 592 | 137 | ||
| AF212320.1 | - | 5 - 6 | 618 | 137 | ||
| AF212321.1 | - | 6 - 7 | 749 | 137 | ||
| AK053850.1 | - | 5 - 6 | 612 | 137 | ||
| AK144608.1 | - | 6 - 7 | 750 | 137 | ||
| AK144745.1 | - | 5 - 6 | 611 | 137 | ||
| AK154929.1 | - | 6 - 7 | 770 | 137 | ||
| AK165897.1 | - | 5 - 6 | 604 | 137 | ||
| AK166084.1 | - | 6 - 7 | 740 | 137 | ||
| AK166094.1 | - | 5 - 6 | 609 | 137 | ||
| AK168020.1 | - | 5 - 6 | 608 | 137 | ||
| AK168025.1 | - | 5 - 6 | 608 | 137 | ||
| AK168170.1 | - | 5 - 6 | 608 | 137 | ||
| AK168679.1 | - | 5 - 6 | 592 | 137 | ||
| AK183940.1 | - | 3 - 4 | 242 | 137 | ||
| AK184356.1 | - | 1 - 2 | 137 | |||
| AK193352.1 | - | 1 - 2 | 81 | 137 | ||
| AK200143.1 | - | 1 - 2 | 81 | 137 | ||
| AK207314.1 | - | 3 - 4 | 242 | 137 | ||
| BC010715.1 | - | 5 - 6 | 597 | 3602242 | 137 | |
More Information
Updated Mapping Information
Set Membership: |
|

Racgap1
Genomic Map