Assay Details
| Gene Symbol: Lemd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146075.2 | NP_666187.2 | 6 - 7 | 1201 | 70 | |
| GenBank mRNA | AK082131.1 | - | 6 - 7 | 1008 | 70 | |
| AK143224.1 | - | 6 - 7 | 1186 | 70 | ||
| AY640307.1 | - | 6 - 7 | 1181 | 70 | ||
| BC026588.1 | - | 6 - 7 | 511 | 4972900 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lemd2
Genomic Map