Assay Details
| Gene Symbol: Smdt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026914.1 | NP_081190.1 | 1 - 2 | 229 | 65 | |
| GenBank mRNA | AK005345.1 | - | 1 - 2 | 221 | 65 | |
| AK170899.1 | - | 1 - 2 | 219 | 65 | ||
| BC028457.1 | - | 1 - 2 | 229 | 1245697 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30382916 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30382913 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30382912 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30382915 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30382917 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P501815 | BC028457.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P501815 | BC092006.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P501815 | NM_026914.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P501815 | AK005345.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P501815 | AK151325.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P501815 | BC028457.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P501815 | BC092006.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P501815 | NM_026914.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P501815 | AK005345.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P501815 | AK151325.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 644760 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P501815 | BC028457.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P501815 | BC092006.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P501815 | NM_026914.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P501815 | AK005345.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P501815 | AK151325.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P501815 | BC028457.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P501815 | BC092006.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P501815 | NM_026914.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P501815 | AK005345.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P501815 | AK151325.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30382927 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30382913 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30382926 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30382931 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30382920 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30382923 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30382922 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30382919 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30382918 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30382915 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30382928 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30382930 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08437853 | - | - |
| Whole Mouse Genome G4122A | A_51_P501815 | BC028457.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P501815 | BC092006.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P501815 | NM_026914.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P501815 | AK005345.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P501815 | AK151325.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P501815 | BC028457.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P501815 | BC092006.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P501815 | NM_026914.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P501815 | AK005345.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P501815 | AK151325.1 | mouse_transcriptome |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12173284 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12173283 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30382912 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30382915 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30382916 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30382917 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30382913 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382996 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382849 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382983 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382978 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382851 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382876 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382987 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382967 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382853 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382985 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382997 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382958 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382931 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382864 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382951 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382966 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382875 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30383003 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382982 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382927 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382877 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382977 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382998 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382972 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382973 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382961 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382869 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382974 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382856 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382980 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382919 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382969 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382993 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382848 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382890 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382923 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382970 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382990 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382854 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382913 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382879 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382922 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382889 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382999 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382920 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382992 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382882 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382956 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382988 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382881 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382925 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382965 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382868 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382888 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382860 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382858 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382991 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382949 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382926 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30383002 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382915 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382918 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382995 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382874 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382938 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382855 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382930 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11897684 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12392228 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12160888 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11897681 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12173282 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30382924 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30382925 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12188797 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30382922 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12160886 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30382931 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12188794 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30382937 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30382939 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12173286 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30382930 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30382913 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30382911 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30382926 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10425759 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10425758 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10425760 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1452584_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10425757 | - | - |
| Mouse Genome 430 2.0 Array | 1452584_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1452584_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 69029_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 69029_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1500005929.mm.1 | NM_026914.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500005929.mm.1 | NM_026914.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500005929.mm.1 | TC1500002336.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500005932.mm.1 | NM_026914.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500005932.mm.1 | NM_026914.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500005932.mm.1 | TC1500002336.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500019342.mm.1 | NM_026914.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500019342.mm.1 | NM_026914.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500019342.mm.1 | TC1500002336.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500005928.mm.1 | NM_026914.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500005928.mm.1 | NM_026914.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500005928.mm.1 | TC1500002336.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500005931.mm.1 | NM_026914.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500005931.mm.1 | NM_026914.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500005931.mm.1 | TC1500002336.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1452584_at | - | - |
| Mouse Expression Set 430 | 1452584_PM_at | - | - |
| Murine Genome U74v2 Set | 95076_PM_at | - | - |
| Murine Genome U74v2 Set | 95076_at | - | - |
| Mouse Genome 430A 2.0 Array | 1452584_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1452584_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1452584_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1452584_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 927369 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 2480242 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2648580 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2648580 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2648580 | - | - |

Smdt1
Genomic Map