Assay Details
| Gene Symbol: Ppm1j | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027982.2 | NP_082258.2 | 4 - 5 | 882 | 79 | |
| GenBank mRNA | AK009235.1 | - | 4 - 5 | 821 | 79 | |
| AY184802.1 | - | 4 - 5 | 882 | 79 | ||
| BC117498.1 | - | 4 - 5 | 878 | 40106734 | 79 | |
| BC121788.1 | - | 4 - 5 | 877 | 40106732 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ppm1j
Genomic Map