Assay Details
| Gene Symbol: Ttc25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028918.2 | NP_083194.2 | 8 - 9 | 1305 | 66 | |
| GenBank mRNA | AK016657.1 | - | 8 - 9 | 1236 | 66 | |
| AK136690.1 | - | 2 - 3 | 150 | 66 | ||
| AK161541.1 | - | 8 - 9 | 1244 | 66 | ||
| AK162855.1 | - | 8 - 9 | 1245 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ttc25
Genomic Map