| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1100013650.mm.1 |
NM_201609.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100013650.mm.1 |
NM_201609.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100013650.mm.1 |
TC1100001469.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100013647.mm.1 |
NM_201609.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100013647.mm.1 |
NM_201609.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100013647.mm.1 |
TC1100001469.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100013646.mm.1 |
NM_201609.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100013646.mm.1 |
NM_201609.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100013646.mm.1 |
TC1100001469.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100013649.mm.1 |
NM_201609.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100013649.mm.1 |
NM_201609.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100013649.mm.1 |
TC1100001469.mm.1 |
MTA TC |