Assay Details
| Gene Symbol: Zfp652 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201609.2 | NP_963903.2 | 3 - 4 | 1341 | 70 | |
| GenBank mRNA | AK220361.1 | - | 3 - 4 | 1186 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Zfp652
Genomic Map