Assay Details
| Gene Symbol: Ube2z | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172300.3 | NP_758504.3 | 5 - 6 | 901 | 77 | |
| GenBank mRNA | AK005524.1 | - | 3 - 4 | 452 | 77 | |
| AK077793.1 | - | 5 - 6 | 900 | 77 | ||
| AK149770.1 | - | 5 - 6 | 901 | 77 | ||
| AK152529.1 | - | 5 - 6 | 615 | 77 | ||
| AK153328.1 | - | 5 - 6 | 632 | 77 | ||
| AK168942.1 | - | 5 - 6 | 594 | 77 | ||
| BC027266.1 | - | 5 - 6 | 526 | 77 | ||
| BC054412.1 | - | 5 - 6 | 604 | 2646768 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ube2z
Genomic Map