| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1700000446.mm.1 |
NM_031395.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700000446.mm.1 |
NM_031395.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700000446.mm.1 |
NM_183370.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700000446.mm.1 |
NM_183370.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700000446.mm.1 |
TC1700000064.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700000449.mm.1 |
NM_183368.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700000449.mm.1 |
NM_183368.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700000449.mm.1 |
TC1700000064.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700000444.mm.1 |
NM_031395.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700000444.mm.1 |
NM_031395.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700000444.mm.1 |
TC1700000064.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700000445.mm.1 |
NM_183370.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700000445.mm.1 |
NM_183370.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700000445.mm.1 |
TC1700000064.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700000450.mm.1 |
NM_031395.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700000450.mm.1 |
NM_031395.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700000450.mm.1 |
NM_183368.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700000450.mm.1 |
NM_183368.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700000450.mm.1 |
NM_183370.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700000450.mm.1 |
NM_183370.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700000450.mm.1 |
TC1700000064.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700000455.mm.1 |
NM_031395.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700000455.mm.1 |
NM_031395.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700000455.mm.1 |
NM_183368.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700000455.mm.1 |
NM_183368.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700000455.mm.1 |
NM_183370.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700000455.mm.1 |
NM_183370.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700000455.mm.1 |
TC1700000064.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700000440.mm.1 |
NM_031395.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700000440.mm.1 |
NM_031395.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700000440.mm.1 |
TC1700000064.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700000448.mm.1 |
NM_031395.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700000448.mm.1 |
NM_031395.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700000448.mm.1 |
NM_183370.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700000448.mm.1 |
NM_183370.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700000448.mm.1 |
TC1700000064.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700000463.mm.1 |
NM_031395.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700000463.mm.1 |
NM_031395.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700000463.mm.1 |
NM_183368.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700000463.mm.1 |
NM_183368.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700000463.mm.1 |
NM_183370.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700000463.mm.1 |
NM_183370.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700000463.mm.1 |
TC1700000064.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700000461.mm.1 |
NM_031395.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700000461.mm.1 |
NM_031395.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700000461.mm.1 |
NM_183368.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700000461.mm.1 |
NM_183368.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700000461.mm.1 |
NM_183370.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700000461.mm.1 |
NM_183370.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700000461.mm.1 |
TC1700000064.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700000439.mm.1 |
NM_031395.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700000439.mm.1 |
NM_031395.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700000439.mm.1 |
TC1700000064.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700000458.mm.1 |
NM_031395.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700000458.mm.1 |
NM_031395.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700000458.mm.1 |
NM_183368.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700000458.mm.1 |
NM_183368.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700000458.mm.1 |
NM_183370.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700000458.mm.1 |
NM_183370.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700000458.mm.1 |
TC1700000064.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700000441.mm.1 |
NM_031395.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700000441.mm.1 |
NM_031395.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700000441.mm.1 |
TC1700000064.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700000460.mm.1 |
NM_031395.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700000460.mm.1 |
NM_031395.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700000460.mm.1 |
NM_183368.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700000460.mm.1 |
NM_183368.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700000460.mm.1 |
NM_183370.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700000460.mm.1 |
NM_183370.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700000460.mm.1 |
TC1700000064.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700000447.mm.1 |
NM_031395.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700000447.mm.1 |
NM_031395.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700000447.mm.1 |
NM_183370.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700000447.mm.1 |
NM_183370.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700000447.mm.1 |
TC1700000064.mm.1 |
MTA TC |