Assay Details
| Gene Symbol: Prr11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175563.5 | NP_780772.1 | 1 - 2 | 76 | 71 | |
| GenBank mRNA | AK040348.1 | - | 1 - 2 | 54 | 71 | |
| AK047463.1 | - | 1 - 2 | 54 | 71 | ||
| AK083651.1 | - | 1 - 2 | 71 | 71 | ||
| BC023918.1 | - | 1 - 2 | 86 | 5323297 | 71 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK040348.1,AK047463.1,AK083651.1,BC023918.1,NM_175563.5 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Prr11
Genomic Map