Assay Details
| Gene Symbol: Serhl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023475.3 | NP_075964.1 | 9 - 10 | 773 | 76 | |
| GenBank mRNA | AJ245737.1 | - | 776 | 76 | ||
| AJ251200.1 | - | 9 - 10 | 776 | 76 | ||
| AK002313.1 | - | 9 - 10 | 686 | 76 | ||
| AK003827.1 | - | 9 - 10 | 686 | 76 | ||
| AK154770.1 | - | 8 - 9 | 1981 | 76 | ||
| BC024048.1 | - | 9 - 10 | 654 | 76 | ||
| BC046342.1 | - | 9 - 10 | 658 | 6475904 | 76 | |
| BC055431.1 | - | 9 - 10 | 785 | 3673247 | 76 | |
| BC064069.1 | - | 7 - 8 | 823 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Serhl
Genomic Map