Assay Details
| Gene Symbol: Vsig1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030181.3 | NP_084457.2 | 2 - 3 | 278 | 99 | |
| GenBank mRNA | AK019565.1 | - | 2 - 3 | 267 | 99 | |
| AK132956.1 | - | 2 - 3 | 279 | 99 | ||
| AK136562.1 | - | 2 - 3 | 262 | 99 | ||
| AK160478.1 | - | 2 - 3 | 265 | 99 | ||
| BC120675.1 | - | 2 - 3 | 234 | 40129598 | 99 | |
| BC125278.1 | - | 2 - 3 | 234 | 40129793 | 99 | |
| DQ007336.1 | - | 2 - 3 | 267 | 99 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Vsig1
Genomic Map