Assay Details
| Gene Symbol: Vsig1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026103.1 | NP_080379.1 | 2 - 3 | 262 | 109 | |
| NM_030181.3 | NP_084457.2 | 3 - 4 | 487 | 109 | ||
| GenBank mRNA | AK006251.1 | - | 2 - 3 | 253 | 109 | |
| AK006862.1 | - | 2 - 3 | 262 | 109 | ||
| AK132956.1 | - | 3 - 4 | 488 | 109 | ||
| AK136562.1 | - | 3 - 4 | 471 | 109 | ||
| AK160478.1 | - | 3 - 4 | 474 | 109 | ||
| BC048589.1 | - | 2 - 3 | 252 | 109 | ||
| BC120675.1 | - | 3 - 4 | 443 | 40129598 | 109 | |
| BC125278.1 | - | 3 - 4 | 443 | 40129793 | 109 | |
| BC145050.1 | - | 4 - 5 | 551 | 9053598 | 109 | |
More Information
Updated Mapping Information
Set Membership: |
|

Vsig1
Genomic Map