Assay Details
| Gene Symbol: Vsig1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026103.1 | NP_080379.1 | 4 - 5 | 540 | 79 | |
| NM_030181.3 | NP_084457.2 | 5 - 6 | 765 | 79 | ||
| GenBank mRNA | AK006862.1 | - | 4 - 5 | 540 | 79 | |
| AK019565.1 | - | 5 - 6 | 754 | 79 | ||
| AK132956.1 | - | 5 - 6 | 766 | 79 | ||
| AK136562.1 | - | 5 - 6 | 749 | 79 | ||
| AK160478.1 | - | 5 - 6 | 752 | 79 | ||
| BC048589.1 | - | 4 - 5 | 530 | 79 | ||
| BC120675.1 | - | 5 - 6 | 721 | 40129598 | 79 | |
| BC125278.1 | - | 5 - 6 | 721 | 40129793 | 79 | |
| BC145050.1 | - | 6 - 7 | 829 | 9053598 | 79 | |
| DQ007336.1 | - | 5 - 6 | 754 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Vsig1
Genomic Map