Assay Details
| Gene Symbol: Ptgr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252625.1 | NP_001239554.1 | 2 - 3 | 322 | 130 | |
| GenBank mRNA | AK145232.1 | - | 2 - 3 | 213 | 130 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK145232.1, NM_001252625.1 transcripts.
Set Membership: |
|

Ptgr2
Genomic Map