Assay Details
| Gene Symbol: Arel1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178065.4 | NP_835166.3 | 6 - 7 | 1285 | 92 | |
| GenBank mRNA | AB093230.1 | - | 5 - 6 | 904 | 92 | |
| AK077015.1 | - | 5 - 6 | 817 | 92 | ||
| AK133332.1 | - | 5 - 6 | 820 | 92 | ||
| AK147491.1 | - | 6 - 7 | 1286 | 92 | ||
| AK147878.1 | - | 6 - 7 | 1249 | 92 | ||
| AK154529.1 | - | 6 - 7 | 1266 | 92 | ||
| BC057351.1 | - | 6 - 7 | 1223 | 92 | ||
| BC060658.1 | - | 5 - 6 | 854 | 30355406 | 92 | |
| BC079590.1 | - | 6 - 7 | 1243 | 92 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Arel1
Genomic Map