Assay Details
| Gene Symbol: Casq2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009814.3 | NP_033944.2 | 7 - 8 | 968 | 67 | |
| GenBank mRNA | AF068244.1 | - | 7 - 8 | 913 | 67 | |
| BC092229.1 | - | 7 - 8 | 881 | 30603069 | 67 | |
| BC140959.1 | - | 7 - 8 | 877 | 9055571 | 67 | |
| U91483.1 | - | 7 - 8 | 787 | 67 | ||
More Information
Set Membership: |
|

Casq2
Genomic Map