Assay Details
| Gene Symbol: Casq2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009814.3 | NP_033944.2 | 8 - 9 | 1022 | 93 | |
| GenBank mRNA | AF068244.1 | - | 8 - 9 | 967 | 93 | |
| BC092229.1 | - | 8 - 9 | 935 | 30603069 | 93 | |
| BC137668.1 | - | 9 - 10 | 940 | 8860687 | 93 | |
| BC140959.1 | - | 8 - 9 | 931 | 9055571 | 93 | |
More Information
Updated Mapping Information
Set Membership: |
|

Casq2
Genomic Map