Assay Details
| Gene Symbol: Arg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009705.3 | NP_033835.1 | 5 - 6 | 696 | 99 | |
| GenBank mRNA | AF032466.1 | - | 5 - 6 | 648 | 99 | |
| BC023349.1 | - | 5 - 6 | 660 | 5254031 | 99 | |
| U90886.1 | - | 5 - 6 | 668 | 99 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Arg2
Genomic Map