Assay Details
| Gene Symbol: Arg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009705.3 | NP_033835.1 | 6 - 7 | 799 | 97 | |
| GenBank mRNA | AF032466.1 | - | 6 - 7 | 751 | 97 | |
| U90886.1 | - | 6 - 7 | 771 | 97 | ||
More Information
Set Membership: |
|

Arg2
Genomic Map