Assay Details
| Gene Symbol: Rab40c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139154.2 | NP_631893.1 | 3 - 4 | 308 | 68 | |
| GenBank mRNA | AB232639.1 | - | 2 - 3 | 202 | 68 | |
| AF422144.1 | - | 2 - 3 | 261 | 68 | ||
| AK133766.1 | - | 2 - 3 | 84 | 68 | ||
| BC027008.1 | - | 3 - 4 | 296 | 5364768 | 68 | |
| BC037461.1 | - | 3 - 4 | 429 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rab40c
Genomic Map