Assay Details
| Gene Symbol: Rab40c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139154.2 | NP_631893.1 | 4 - 5 | 366 | 71 | |
| GenBank mRNA | AB232639.1 | - | 3 - 4 | 260 | 71 | |
| AF422144.1 | - | 3 - 4 | 319 | 71 | ||
| AK133766.1 | - | 3 - 4 | 142 | 71 | ||
| BC027008.1 | - | 4 - 5 | 354 | 5364768 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rab40c
Genomic Map