Assay Details
| Gene Symbol: Miox | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019977.2 | NP_064361.2 | 6 - 7 | 551 | 85 | |
| GenBank mRNA | AF197127.1 | - | 6 - 7 | 549 | 85 | |
| AK002406.1 | - | 6 - 7 | 564 | 85 | ||
| AY738257.1 | - | 5 - 6 | 521 | 85 | ||
| BC013543.1 | - | 6 - 7 | 551 | 4242159 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2023200 | BC013543.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2023200 | AK165481.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2023200 | AK002406.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2023200 | NM_019977.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 84166 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2023200 | BC013543.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2023200 | AK165481.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2023200 | AK002406.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2023200 | NM_019977.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P385258 | AF197127.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30424608 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30424602 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30424579 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30424611 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30424600 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30424621 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30424617 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30424625 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30424622 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30424627 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30424618 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30424628 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30424623 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30424612 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30424614 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30424624 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30424607 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30424613 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30424574 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30424605 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30424616 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30424626 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30424619 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30424609 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30424577 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30424603 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30424597 | - | - |
| Whole Mouse Genome G4122A | A_51_P385258 | AF197127.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P109636 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12296047 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12296051 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12296045 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30424611 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30424600 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30424650 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30424616 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30424614 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30424602 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30424649 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30424622 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30424647 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30424627 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30424619 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30424612 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30424590 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30424617 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30424608 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30424623 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30424593 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30424583 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30424624 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30424637 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30424655 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30424652 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30424629 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30424574 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30424577 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30424589 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30424621 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30424584 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30424609 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30424633 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12450434 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12260171 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12296043 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30424617 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30424597 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30424590 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30424608 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12260179 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30424591 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30424579 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11946384 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30424613 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30424612 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30424596 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12296021 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12404249 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30424614 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30424638 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12296050 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30424624 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30424593 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30424600 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30424641 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12423565 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30424607 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12422823 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30424595 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30424605 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30424635 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30424601 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30424602 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30424583 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30424623 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12369267 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30424574 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30424598 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30424627 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12296048 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30424633 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30424609 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30424619 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12016077 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30424637 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30424639 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30424584 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30424592 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30424603 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30424611 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10426220 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10426223 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10426219 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10426221 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10426214 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10426215 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10426216 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10426218 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10426222 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10426224 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1416460_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10426213 | - | - |
| Mouse Genome 430 2.0 Array | 1416460_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1416460_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 56727_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 56727_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1500006842.mm.1 | NM_019977.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500006842.mm.1 | NM_019977.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500006842.mm.1 | TC1500000870.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1416460_PM_at | - | - |
| Mouse Expression Set 430 | 1416460_at | - | - |
| Murine Genome U74v2 Set | 93781_PM_at | - | - |
| Murine Genome U74v2 Set | 93781_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416460_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416460_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416460_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416460_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 4810364 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004810364 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2640116 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2640116 | - | - |
| mouse-6 v1.1 (Illumina) | scl47618.10.1_16-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4810364 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl47618.10.1_16-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4810364 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2640116 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2640116 | - | - |

Miox
Genomic Map