Assay Details
| Gene Symbol: Lyrm9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076681.2 | NP_001070149.1 | 1 - 2 | 46 | 96 | |
| GenBank mRNA | AK007467.1 | - | 1 - 2 | 60 | 96 | |
| AK009070.1 | - | 1 - 2 | 29 | 96 | ||
| AK144380.1 | - | 1 - 2 | 47 | 96 | ||
| BC130224.1 | - | 1 - 2 | 37 | 40141726 | 96 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK144380.1, BC130224.1, NM_001076681.2 transcripts.
Set Membership: |
|

Lyrm9
Genomic Map