Assay Details
| Gene Symbol: Lyrm9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076681.2 | NP_001070149.1 | 3 - 4 | 271 | 94 | |
| GenBank mRNA | AK007467.1 | - | 3 - 4 | 285 | 94 | |
| AK009070.1 | - | 3 - 4 | 254 | 94 | ||
| AK144380.1 | - | 3 - 4 | 272 | 94 | ||
| BC130224.1 | - | 3 - 4 | 262 | 40141726 | 94 | |
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27945541 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27945544 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27945545 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27945543 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27945542 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P393120 | BC031726.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P393120 | NM_001076681.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P393120 | BC031726.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P393120 | NM_001076681.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 261775 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 813038 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 923925 | AF139659.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 923925 | AK078405.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 923925 | AK163256.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 923925 | BC048689.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 923925 | BC080739.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 923925 | NM_019716.1 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 819517 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 736153 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 571787 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 753189 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 139918 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 413459 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 859350 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 51594 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P138970 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P393120 | BC031726.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P393120 | NM_001076681.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P393120 | BC031726.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P393120 | NM_001076681.2 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P138970 | - | - |
| Whole Mouse Genome G4122A | A_52_P540462 | BC130224.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P540462 | NM_001076681.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P540462 | AK144380.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P393120 | BC031726.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P393120 | NM_001076681.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P393120 | BC031726.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P393120 | NM_001076681.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P134146 | BC130224.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P134146 | NM_001076681.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P134146 | AK007467.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P134146 | AK009070.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P134146 | AK144380.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P100129 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P138970 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27945541 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27945542 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27945545 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27945544 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27945543 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27945656 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27945643 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27945634 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27945683 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27945561 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27945632 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27945575 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27945556 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27945532 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27945495 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27945654 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27945562 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27945536 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27945599 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27945507 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27945601 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27945621 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27945691 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27945566 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27945557 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27945510 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27945519 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27945548 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27945570 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27945628 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27945577 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27945535 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27945521 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27945616 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27945530 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27945642 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27945537 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27945639 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27945692 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27945504 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27945655 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27945609 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27945486 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27945622 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27945538 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27945669 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27945607 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27945677 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27945604 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27945680 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27945666 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27945608 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27945485 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27945663 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27945620 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27945679 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27945572 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27945613 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27945568 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27945695 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27945492 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27945584 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27945503 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27945672 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27945524 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27945661 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27945619 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27945478 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27945657 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27945487 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27945658 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27945483 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27945583 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27945647 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27945502 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27945513 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27945653 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27945505 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27945586 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27945559 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27945490 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27945516 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27945595 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27945543 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27945618 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27945592 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27945541 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27945650 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27945544 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27945491 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27945481 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27945675 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27945670 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27945501 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27945651 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27945554 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27945546 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27945610 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27945635 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27945640 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27945631 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27945482 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27945685 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27945681 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27945688 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27945571 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27945550 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27945558 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27945544 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27945546 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27945535 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27945482 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27945519 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27945496 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27945588 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27945570 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27945575 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27945495 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09616862 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27945505 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27945478 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09155457 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27945586 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27945481 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27945532 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27945511 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09155463 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27945581 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09348926 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27945583 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27945537 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27945559 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27945503 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08994957 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27945526 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27945556 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27945545 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09297672 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27945516 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27945491 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27945521 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27945524 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27945490 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27945509 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27945513 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27945499 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27945510 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27945501 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09297670 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27945589 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27945489 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27945548 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09574529 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27945487 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27945554 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27945550 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09399142 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09355296 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27945504 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27945568 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27945572 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27945530 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27945485 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27945486 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09042493 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27945585 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08994958 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09155448 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27945571 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27945492 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09522890 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27945497 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27945483 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27945562 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27945502 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10379225 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10379227 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10379226 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10379224 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1458028_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1436548_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1440234_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10379223 | - | - |
| Mouse Genome 430 2.0 Array | 1440234_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1436548_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1440234_at | - | - |
| Mouse Genome 430 2.0 Array | 1436548_at | - | - |
| Mouse Genome 430 2.0 Array | 1458028_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1458028_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66274_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66274_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1100010905.mm.1 | NM_001076681.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100010905.mm.1 | NM_001076681.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100010905.mm.1 | TC1100001169.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100010907.mm.1 | NM_001076681.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100010907.mm.1 | NM_001076681.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100010907.mm.1 | TC1100001169.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100010906.mm.1 | NM_001076681.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100010906.mm.1 | NM_001076681.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100010906.mm.1 | TC1100001169.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1436548_at | - | - |
| Mouse Expression Set 430 | 1458028_PM_at | - | - |
| Mouse Expression Set 430 | 1436548_PM_at | - | - |
| Mouse Expression Set 430 | 1440234_at | - | - |
| Mouse Expression Set 430 | 1458028_at | - | - |
| Mouse Expression Set 430 | 1440234_PM_at | - | - |
| Murine Genome U74v2 Set | 130033_PM_at | - | - |
| Murine Genome U74v2 Set | 130033_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1458028_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1440234_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1440234_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1436548_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1436548_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1458028_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 940164 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1229428 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1229428 | - | - |

Lyrm9
Genomic Map