Assay Details
| Gene Symbol: Krt19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008471.2 | NP_032497.1 | 4 - 5 | 889 | 65 | |
| GenBank mRNA | AB472075.1 | - | 4 - 5 | 889 | 65 | |
| BC034561.1 | - | 4 - 5 | 882 | 2655016 | 65 | |
| BC085304.1 | - | 4 - 5 | 889 | 6437375 | 65 | |
| M28698.1 | - | 4 - 5 | 884 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Krt19
Genomic Map