Assay Details
| Gene Symbol: Igsf23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027308.2 | NP_081584.1 | 3 - 4 | 782 | 62 | |
| GenBank mRNA | AK033677.1 | - | 3 - 4 | 783 | 62 | |
| AK088927.1 | - | 4 - 5 | 738 | 62 | ||
| AK135657.1 | - | 588 | 62 | |||
| BC139455.1 | - | 3 - 4 | 630 | 8862477 | 62 | |
| BC139458.1 | - | 3 - 4 | 630 | 8862480 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Igsf23
Genomic Map