Assay Details
| Gene Symbol: Tspan2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027533.3 | NP_081809.2 | 1 - 2 | 297 | 78 | |
| GenBank mRNA | AK018178.1 | - | 1 - 2 | 100 | 78 | |
| AK032520.1 | - | 1 - 2 | 123 | 78 | ||
| AK049888.1 | - | 1 - 2 | 99 | 78 | ||
| AK079560.1 | - | 1 - 2 | 101 | 78 | ||
| BB642061.1 | - | 1 - 2 | 298 | 78 | ||
| BC007185.1 | - | 1 - 2 | 538 | 3591472 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tspan2
Genomic Map