Assay Details
| Gene Symbol: Mill1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153749.4 | NP_715630.3 | 1 - 2 | 354 | 66 | |
| GenBank mRNA | AB086265.1 | - | 1 - 2 | 119 | 66 | |
| AK029010.1 | - | 1 - 2 | 355 | 66 | ||
| AK030695.1 | - | 1 - 2 | 278 | 66 | ||
| BC110547.1 | - | 1 - 2 | 330 | 40047031 | 66 | |
| BC110548.1 | - | 1 - 2 | 330 | 40047032 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mill1
Genomic Map