Assay Details
| Gene Symbol: Epx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007946.2 | NP_031972.2 | 9 - 10 | 1662 | 62 | |
| GenBank mRNA | AK132416.1 | - | 4 - 5 | 787 | 62 | |
| BC052881.1 | - | 9 - 10 | 1644 | 62 | ||
| L77979.1 | - | 9 - 10 | 1646 | 62 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Epx
Genomic Map