Assay Details
| Gene Symbol: Brd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033274.3 | NP_001028446.2 | 3 - 4 | 1830 | 63 | |
| GenBank mRNA | AK149714.1 | - | 3 - 4 | 1842 | 63 | |
| AK220233.1 | - | 2 - 3 | 751 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Brd1
Genomic Map