Assay Details
| Gene Symbol: Brd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033274.3 | NP_001028446.2 | 5 - 6 | 2095 | 84 | |
| GenBank mRNA | AK149714.1 | - | 5 - 6 | 2107 | 84 | |
| AK220233.1 | - | 4 - 5 | 1016 | 84 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Brd1
Genomic Map