Assay Details
| Gene Symbol: Brd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK149714.1 | - | 7 - 8 | 2674 | 87 | |
| BC037388.1 | - | 2 - 3 | 382 | 87 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Brd1
Genomic Map