Assay Details
| Gene Symbol: 9430007A20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198662.3 | NP_941064.2 | 2 - 3 | 446 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

9430007A20Rik| Gene Symbol: 9430007A20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198662.3 | NP_941064.2 | 2 - 3 | 446 | 63 | |
Set Membership: |
|