Assay Details
| Gene Symbol: Cox16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025461.5 | NP_079737.1 | 2 - 3 | 289 | 138 | |
| GenBank mRNA | AK007450.1 | - | 2 - 3 | 266 | 138 | |
| AK007562.1 | - | 2 - 3 | 278 | 138 | ||
| AK008486.1 | - | 2 - 3 | 254 | 138 | ||
| AK040003.1 | - | 2 - 3 | 291 | 138 | ||
| AK082374.1 | - | 2 - 3 | 290 | 138 | ||
More Information
Set Membership: |
|

Cox16
Genomic Map