Assay Details
| Gene Symbol: Man1a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010763.2 | NP_034893.1 | 8 - 9 | 1781 | 124 | |
| GenBank mRNA | AK153336.1 | - | 8 - 9 | 1781 | 124 | |
| BC049121.1 | - | 8 - 9 | 1185 | 6813258 | 124 | |
| BC068192.1 | - | 8 - 9 | 1185 | 6813258 | 124 | |
| U03458.1 | - | 8 - 9 | 1759 | 124 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Man1a2
Genomic Map