Assay Details
| Gene Symbol: Ccnt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009833.1 | NP_033963.1 | 1 - 2 | 160 | 118 | |
| GenBank mRNA | AF087662.1 | - | 1 - 2 | 160 | 118 | |
| AF095640.1 | - | 1 - 2 | 160 | 118 | ||
| AF113951.1 | - | 1 - 2 | 160 | 118 | ||
| AK083037.1 | - | 1 - 2 | 204 | 118 | ||
| AK133168.1 | - | 860 | 118 | |||
| BC131685.1 | - | 1 - 2 | 160 | 40111509 | 118 | |
| BC131686.1 | - | 1 - 2 | 160 | 118 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ccnt1
Genomic Map