Assay Details
| Gene Symbol: Mrpl28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024227.3 | NP_077189.2 | 3 - 4 | 635 | 125 | |
| GenBank mRNA | AK003727.2 | - | 3 - 4 | 616 | 125 | |
| AK031701.1 | - | 3 - 4 | 605 | 125 | ||
| AK046544.1 | - | 3 - 4 | 616 | 125 | ||
| AK163190.1 | - | 3 - 4 | 618 | 125 | ||
| BC005797.1 | - | 3 - 4 | 607 | 125 | ||
| BC080753.1 | - | 3 - 4 | 581 | 125 | ||
| BC100322.1 | - | 3 - 4 | 607 | 125 | ||
| BC145522.1 | - | 3 - 4 | 614 | 125 | ||
| BC145680.1 | - | 3 - 4 | 604 | 40130983 | 125 | |
| BC145682.1 | - | 3 - 4 | 604 | 40130985 | 125 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mrpl28
Genomic Map