Assay Details
| Gene Symbol: Rgs11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081069.1 | NP_001074538.1 | 13 - 14 | 995 | 68 | |
| GenBank mRNA | AF061934.1 | - | 12 - 13 | 900 | 68 | |
| AK039314.1 | - | 12 - 13 | 1617 | 68 | ||
| AK080707.1 | - | 13 - 14 | 1087 | 68 | ||
| AK161551.1 | - | 9 - 10 | 1054 | 68 | ||
| BC019741.1 | - | 2 - 3 | 327 | 68 | ||
| BC049910.1 | - | 11 - 12 | 1699 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rgs11
Genomic Map