Assay Details
| Gene Symbol: Rgs11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081069.1 | NP_001074538.1 | 14 - 15 | 1076 | 110 | |
| GenBank mRNA | AF061934.1 | - | 13 - 14 | 981 | 110 | |
| AK039314.1 | - | 13 - 14 | 1698 | 110 | ||
| AK080707.1 | - | 14 - 15 | 1170 | 112 | ||
| AK161551.1 | - | 10 - 11 | 1135 | 110 | ||
| BC019741.1 | - | 3 - 4 | 408 | 110 | ||
| BC049910.1 | - | 12 - 13 | 1780 | 110 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rgs11
Genomic Map