Assay Details
| Gene Symbol: Rgs11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081069.1 | NP_001074538.1 | 15 - 16 | 1210 | 66 | |
| GenBank mRNA | AF061934.1 | - | 14 - 15 | 1115 | 66 | |
| AK039314.1 | - | 14 - 15 | 1832 | 66 | ||
| AK080707.1 | - | 15 - 16 | 1304 | 66 | ||
| AK161551.1 | - | 11 - 12 | 1269 | 66 | ||
| AK166716.1 | - | 1 - 2 | 455 | 66 | ||
| BC019741.1 | - | 4 - 5 | 542 | 66 | ||
| BC049910.1 | - | 13 - 14 | 1914 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
Function(s)
Process(es)
G-protein coupled receptor signaling pathway
negative regulation of signal transduction
intracellular signal transduction
termination of G-protein coupled receptor signaling pathway
positive regulation of GTPase activity
signal transducer activity
GTPase activator activity
protein binding
G-protein beta-subunit binding
G-protein coupled receptor signaling pathway
negative regulation of signal transduction
intracellular signal transduction
termination of G-protein coupled receptor signaling pathway
positive regulation of GTPase activity
signal transducer activity
GTPase activator activity
protein binding
G-protein beta-subunit binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2000354 | BC049910.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2000354 | AK080707.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2000354 | AK161551.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2000354 | AK166716.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2000354 | BC019741.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 887274 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 235142 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 859604 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 927887 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 212799 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P139767 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2000354 | BC049910.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2000354 | AK080707.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2000354 | AK161551.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2000354 | AK166716.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2000354 | BC019741.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P139767 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08744864 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P139767 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108928 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108878 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108909 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108907 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108888 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108861 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108957 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108915 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108921 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108929 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108884 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108963 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108880 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108855 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108879 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108977 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108865 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108854 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108968 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108895 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108976 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108870 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108953 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108908 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108872 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108892 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108885 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108935 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108904 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108902 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108857 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108913 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108938 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108905 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108859 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108933 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108973 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108972 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108918 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108910 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108947 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108967 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108969 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108955 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108926 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108882 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108942 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108877 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108891 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108924 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108951 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108934 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108874 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108959 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108875 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108886 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108868 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108903 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108970 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31108929 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13254111 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31108951 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31108953 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31108947 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31108955 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31108975 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31108907 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13317398 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31108909 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31108957 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31108945 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31108908 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31108959 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31108944 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31108946 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31108934 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13521531 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13323073 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31108935 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31108918 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31108928 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31108921 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31108965 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31108969 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31108961 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13323069 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13323078 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31108973 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31108933 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31108910 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13073559 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31108923 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31108924 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13323066 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13323065 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31108976 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31108942 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13317410 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31108938 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13254109 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10442970 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10442978 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10442976 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10442983 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10442972 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10442977 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10442971 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10442973 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10442974 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10442982 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10442969 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10442975 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10442984 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10442980 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10442981 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10442985 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1425245_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10442968 | - | - |
| Mouse Genome 430 2.0 Array | 1425245_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1425245_PM_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 50782_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 50782_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1700003467.mm.1 | NM_001081069.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700003467.mm.1 | NM_001081069.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700003467.mm.1 | TC1700000407.mm.1 | MTA TC |
| Murine Genome U74v2 Set | 102650_at | - | - |
| Murine Genome U74v2 Set | 161309_r_at | - | - |
| Murine Genome U74v2 Set | 102650_PM_at | - | - |
| Murine Genome U74v2 Set | 161309_PM_r_at | - | - |
| Murine Genome U74v2 Set | 169368_PM_at | - | - |
| Murine Genome U74v2 Set | 169368_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1425245_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1425245_PM_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 439922 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2667203 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2667203 | - | - |

Rgs11
Genomic Map