Assay Details
| Gene Symbol: Rgs11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081069.1 | NP_001074538.1 | 4 - 5 | 334 | 79 | |
| GenBank mRNA | AK080707.1 | - | 4 - 5 | 424 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rgs11
Genomic Map