Assay Details
| Gene Symbol: Rgs11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081069.1 | NP_001074538.1 | 9 - 10 | 675 | 67 | |
| GenBank mRNA | AF061934.1 | - | 8 - 9 | 580 | 67 | |
| AK080707.1 | - | 9 - 10 | 766 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rgs11
Genomic Map