Assay Details
| Gene Symbol: Poldip2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026389.3 | NP_080665.1 | 8 - 9 | 809 | 71 | |
| GenBank mRNA | AK003702.1 | - | 6 - 7 | 475 | 71 | |
| BC006833.1 | - | 8 - 9 | 809 | 3597851 | 71 | |
| BC012879.1 | - | 8 - 9 | 821 | 4166791 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Poldip2
Genomic Map