Assay Details
| Gene Symbol: Lgals9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010708.2 | NP_034838.2 | 4 - 5 | 547 | 73 | |
| GenBank mRNA | CA458877.1 | - | 4 - 5 | 537 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lgals9
Genomic Map