Assay Details
| Gene Symbol: Tnfaip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159392.1 | NP_001152864.1 | 2 - 3 | 495 | 62 | |
| NM_009395.4 | NP_033421.3 | 2 - 3 | 508 | 62 | ||
| GenBank mRNA | AK004593.2 | - | 2 - 3 | 422 | 62 | |
| AK034305.1 | - | 2 - 3 | 443 | 62 | ||
| AK147809.1 | - | 2 - 3 | 403 | 62 | ||
| AK168386.1 | - | 2 - 3 | 387 | 62 | ||
| AK168704.1 | - | 2 - 3 | 430 | 62 | ||
| AK168805.1 | - | 2 - 3 | 403 | 62 | ||
| BC003906.1 | - | 2 - 3 | 344 | 3484017 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tnfaip1
Genomic Map