Assay Details
| Gene Symbol: Tnfaip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159392.1 | NP_001152864.1 | 3 - 4 | 664 | 55 | |
| NM_009395.4 | NP_033421.3 | 3 - 4 | 677 | 55 | ||
| GenBank mRNA | AK004593.2 | - | 3 - 4 | 591 | 55 | |
| AK034305.1 | - | 3 - 4 | 612 | 55 | ||
| AK147809.1 | - | 3 - 4 | 572 | 55 | ||
| AK168386.1 | - | 3 - 4 | 556 | 55 | ||
| AK168704.1 | - | 3 - 4 | 599 | 55 | ||
| AK168805.1 | - | 3 - 4 | 572 | 55 | ||
| AK216341.1 | - | 2 - 3 | 234 | 55 | ||
| BC003906.1 | - | 3 - 4 | 513 | 3484017 | 55 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tnfaip1
Genomic Map