Assay Details
| Gene Symbol: Rnf128 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001254761.1 | NP_001241690.1 | 4 - 5 | 946 | 84 | |
| NM_023270.5 | NP_075759.3 | 4 - 5 | 1072 | 84 | ||
| GenBank mRNA | AB041548.1 | - | 4 - 5 | 1072 | 84 | |
| AF426411.1 | - | 4 - 5 | 886 | 84 | ||
| AK004847.1 | - | 4 - 5 | 1073 | 84 | ||
| AK018582.1 | - | 4 - 5 | 1074 | 84 | ||
| AK146266.1 | - | 4 - 5 | 1069 | 84 | ||
| AK167031.1 | - | 4 - 5 | 1058 | 84 | ||
| AY112656.1 | - | 4 - 5 | 1059 | 84 | ||
| BC010477.1 | - | 4 - 5 | 1061 | 3154433 | 84 | |
| DV064262.1 | - | 4 - 5 | 709 | 84 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rnf128
Genomic Map