Assay Details
| Gene Symbol: Oxct1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024188.6 | NP_077150.1 | 10 - 11 | 1398 | 95 | |
| GenBank mRNA | AB085609.1 | - | 10 - 11 | 1140 | 95 | |
| AK009027.1 | - | 10 - 11 | 1178 | 95 | ||
| AK011354.1 | - | 10 - 11 | 1078 | 95 | ||
| AK078086.1 | - | 10 - 11 | 1396 | 95 | ||
| AK146158.1 | - | 10 - 11 | 1156 | 95 | ||
| AK146342.1 | - | 10 - 11 | 1143 | 95 | ||
| AK151662.1 | - | 10 - 11 | 1137 | 95 | ||
| AK151695.1 | - | 10 - 11 | 1128 | 95 | ||
| AK167681.1 | - | 10 - 11 | 1228 | 95 | ||
| AK215681.1 | - | 3 - 4 | 254 | 95 | ||
| BC003422.1 | - | 10 - 11 | 1102 | 2598824 | 95 | |
More Information
Updated Mapping Information
Set Membership: |
|

Oxct1
Genomic Map