Assay Details
| Gene Symbol: Oxct1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024188.6 | NP_077150.1 | 13 - 14 | 1593 | 61 | |
| GenBank mRNA | AB085609.1 | - | 13 - 14 | 1335 | 61 | |
| AK009027.1 | - | 13 - 14 | 1373 | 61 | ||
| AK011354.1 | - | 13 - 14 | 1273 | 61 | ||
| AK078086.1 | - | 13 - 14 | 1591 | 61 | ||
| AK146158.1 | - | 13 - 14 | 1351 | 61 | ||
| AK146342.1 | - | 13 - 14 | 1338 | 61 | ||
| AK151662.1 | - | 13 - 14 | 1332 | 61 | ||
| AK151695.1 | - | 13 - 14 | 1323 | 61 | ||
| AK167681.1 | - | 13 - 14 | 1423 | 61 | ||
| AK185909.1 | - | 3 - 4 | 168 | 61 | ||
| AK217531.1 | - | 3 - 4 | 168 | 61 | ||
| BC003422.1 | - | 13 - 14 | 1297 | 2598824 | 61 | |
More Information
Updated Mapping Information
Set Membership: |
|

Oxct1
Genomic Map